missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Clear Standard Wide Mouth Bottles without Cap

Product Code. 11593672 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
120 mL
240 mL
480 mL
960 mL
3785 mL
Closure Size:
48-400 mm
58-400 mm
63-400 mm
70-400 mm
89-400 mm
Quantity:
12/Cs.
24/Cs.
4/Cs.
Unit Size:
Pack of 12
Pack of 24
Pack of 4
This item is not returnable. View return policy

Product Code. 11593672

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

For pharmaceutical, chemical, environmental applications. Fisherbrand™ Clear Standard Wide Mouth Bottles without Cap are made of glass.

Clear / Flint Standard Wide Mouth Bottles feature a wide opening making them the ideal general purpose bottle. Applications include pharmaceutical, chemical, environmental, and food packaging and storing.

  • Used for food packaging and storing
  • Feature wide opening making them ideal general purpose bottle

Specifications

Capacity (Metric) 960 mL
Capacity (English) 32 oz.
Color Clear
Material Glass
Quantity 12/Cs.
Neck Finish 70-400
Closure Included No
Mouth Wide
Shape Round
Product Type Standard Wide Mouth Bottle
Closure Size 70-400 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.