Learn More
Gibco™ RPMI 1640
Product Code. 16239961
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 16239961
Brand: Gibco™ 670089
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Media with increased levels of vitamins
Contains increased levels of vitamins, the reducing agent glutathione, phenol red, 2 g/L glucose, 2 mM L-glutamine, and 2 g/L sodium bicarbonate. RPMI 1640 does not contain HEPES or sodium pyruvate.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Protect from light, 2°C to 8°C
Product Type Basal Media
For Use With (Application) Cell Culture
Quantity 500 mL