missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RPS6KA2 (RSK3), GST Tag Recombinant Protein

Human RPS6KA2 (RSK3), GST Tag Recombinant Protein

632.00€ - 1826.00€

Specifications

Accession Number NP_066958.2
For Use With (Application) Kinase Assay
KinaseFamily RSK (RPS6K) Kinases
Protein Length Full Length
Species Human
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15512165
View Documents Promotion Details
Invitrogen™
A31517
10μg
632.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15667888
View Documents Promotion Details
Invitrogen™
A31518
100μg
1826.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ Assay technologies to identify novel kinase inhibitors. RPS6KA2 is also available in our SelectScreen™ Kinase Profiling Service.

  • Validated for use with the LanthaScreen & Z'-LYTE assay technology
  • Available in multiple pack sizes for convenient use
  • Also available in our SelectSceen Kinase Profiling Service
Specifications

Specifications

NP_066958.2
RSK (RPS6K) Kinases
Human
GST Tag
Kinase Assay
Full Length
Recombinant
Insect
Videos
SDS
Documents

Documents

Product Certifications