missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPS6KA4 (MSK2) Recombinant Human Protein

RPS6KA4 (MSK2) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes.

1826.00€ - 7890.00€

Specifications

Accession Number NP_003933
For Use With (Application) Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
Formulation 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Molecular Weight (g/mol) 113.1kDa
Storage Requirements Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15488523
View Documents Promotion Details
Invitrogen™
PR7155A
100μg
1826.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10261403
View Documents Promotion Details
Invitrogen™
PV4052
1mg
7890.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 16-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The RPS6KA4 (MSK2) kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos.

Specifications

Specifications

NP_003933
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
RSK (RPS6K) Kinase Family
Recombinant
Baculovirus
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
113.1kDa
RUO - research use only
RPS6KA4 (MSK2)
Human
GST Tag
Frozen
Videos
SDS
Documents

Documents

Product Certifications