missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS6KB1 (p70S6K) Recombinant Human Protein
RPS6KB1 (p70S6K) is a serine/threonine kinase with 2 non-identical kinase catalytic domains.
Brand: Invitrogen™ PV4054
Additional Details : Weight : 0.00900kg
Description
RPS6KB1 (p70S6K) phosphorylates several residues of the S6 ribosomal protein.
Specifications
NP_003152,T412E | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
RPS6KB1 (p70S6K) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
75.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Baculovirus |