Learn More
Abnova™ RSAFD1 Recombinant Protein
Product Code. 16101343
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16101343
Brand: Abnova™ H00055253P01.10ug
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Accession Number AAH51888
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 55253
Molecular Weight (g/mol) 67.98
Name RSAFD1 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MDPSADTWDLFSPLISLWINRFYIYLGFAVSISLWICVQIVIKTQGKNLQEKSVPKAAQDLMTNGYVSLQEKDIFVSGVKIFYGSQTGTAKGFATVLAEAVTSLDLPVAIINLKEYDPDDHLIEEVTSKNVCVFLVATYTDGLPTESAEWFCKWLEEASIDFRFGKTYLKGMRYAVFGLGNSAYASHFNKVGKNVDKWLWMLGAHRVMSRGEGDCDVVKSKHGSIEADFRAWKTKFISQLQALQKGERKKSCGGHCKKGKCESHQHGSEEREEGSHEQDELHHRDTEEEEPFESSSEEEFGGEDHQSLNSIVDVEDLGKIMDHVKKEKREKEQQEEKSGLFRNMGRNEDGERRAMITPALREALTKQVHAPRERRLLQTHILWN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias FLJ10900/MGC23001/MGC60291/RSAFD1/TYW1A/YPL207W
Common Name TYW1
Gene Symbol TYW1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution