Learn More
IKA™ RT 10 Magnetic Stirring Hotplate
GREENER_CHOICE
Product Code. p-7163865 Shop All IKA Products
Click to view available options
Plug Type:
EU
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 13410557
Brand: IKA™ 0003691100
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
10-position digital magnetic hotplate stirrer for synchronous heating and stirring. IKA™ Stainless Steel Digital Magnetic Stirrer provides noiseless and consistent stirring on all positions.
  • Speed remains constant, even when load changes.
  • Homogeneous temperature distribution across the heating plate
  • Adjustable reverse operation
  • Eco-mode for a low self-warming of the surface
  • Wear free magnetic coils
  • Error code display
  • Easy operation with touch keypad
  • Suitable for continuous operation
  • Speed range: 0 to 1000 rpm
TRUSTED_SUSTAINABILITY
Specifications
Speed 0 to 1000 rpm
Stirring Positions 10
Voltage 100/115/230 V
Wattage 410 w
Width (Metric) 190 mm
Electrical Requirements 230/115/100V, 50/60Hz, 410w
Frequency 50/60 Hz
Plug Type EU
Speed Control 10 RPM Steps
Temperature Range (Metric) 5°C to 40°C
Type RT 10 Magnetic Stirrer