Learn More
Abeba™ Rubber 5150 Shoes
ESD dissipative shoe is acid and base resistant.
Brand:  Abeba™ 5150/41
Additional Details : Poids : 0.95000kg
 View more versions of this product
Product Code. 15661828
  • 50.40€ / 1 pair
Expédition estimée
23-09-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Leather top layer
  • Adjustable instep area
  • Lining microfiber, respiration absorbing
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Microfiber
41
Rubber
Blue
Leather
Slip-resistant
5150