Learn More
Abeba™ Rubber 5250 Shoes
ESD dissipative shoe is acid and base resistant.
Brand: Abeba™ 5250/44
60.89 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Dark blue
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Leather top layer
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Leather | |
Slip-resistant | |
5250 |
Blue | |
44 | |
Rubber |