Learn More
Abeba™ Rubber 5250 Shoes
Product Code. 15386711
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15386711 45 1 pair
15621888 36 1 pair
15376701 37 1 pair
15396701 38 1 pair
15306711 39 1 pair
15316711 40 1 pair
15336711 41 1 pair
15346711 42 1 pair
15356711 43 1 pair
15366711 44 1 pair
15312345 46 1 pair
15322345 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15386711 Supplier Abeba™ Supplier No. 5250/45
Only null left
Add to basket
Add to basket
ESD dissipative shoe is acid and base resistant.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color Blue
Material Leather
Size 45
Sole Features Slip-resistant
Sole Material Rubber
Style 5250