Learn More
Abeba™ Rubber 9100 Shoes
Acid and base resistant. Rubber sole provides high slip resistance.
Brand: Abeba™ 9100/45
Additional Details : Weight : 0.95000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- White
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Leather insole
- adjustable instep area
Specifications
CE EN ISO 20347:2012 OB A, FO SRC | |
Leather | |
Slip-resistant | |
9100 |
White | |
45 | |
Rubber |