Learn More
Abeba™ Rubber 9300 Shoes
Product Code. 15623228
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15623228 36 1 pair
15633228 37 1 pair
15643228 38 1 pair
15653228 39 1 pair
15663228 40 1 pair
15673228 41 1 pair
15683228 42 1 pair
15693228 43 1 pair
15603238 44 1 pair
15613238 45 1 pair
15623238 46 1 pair
15633238 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15623228 Supplier Abeba™ Supplier No. 9300/36
Only null left
Add to basket
Add to basket
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • White
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC/li
Color White
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Rubber
Style 9300