Learn More
OHAUS™ Rubber Mat
Shop All Ohaus Products
Click to view available options
Dimensions (L x W):
28 x 33 cm
33 x 33 cm
45.7 x 45.7 cm
48 x 61 cm
61 x 61 cm
61 x 91.4 cm
Unit Size:
Each
Description
- For use with platforms
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Securely hold labware in place
Specifications
Specifications
| Product Type | Platform Mat |
| Material | Rubber |
| Depth (English) | 13.00 in. |
| Depth (Metric) | 33.00 cm |
| Dimensions (L x W) | 33 x 33 cm |
| Length (English) | 13.00 in. |
| Length (Metric) | 33.00 cm |
| Width (Metric) | 33.00 cm |
| Description | Rubber Mat, 33 x 33 cm |
| For Use With (Equipment) | Platform |