missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium solution for ICP/MS 100mg/L HNO3 matrix, Fisher Chemical™
Shop All Fisher Chemical Products

Specifications
Specifications
| Packaging | Bottle |
| CAS | 7697-37-2 |
| Molecular Formula | HNO3 |
| Concentration | 100 mg/L |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| SMILES | O[N+]([O-])=O |
| IUPAC Name | nitric acid |
| Solvent or Matrix | Nitric Acid |
| Molecular Weight (g/mol) | 63.01 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?