Learn More
Applied Biosystems™ 3730 Running Buffer (10X)
Shop All Applied Biosystems Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Buffer | Running Buffer |
| Concentration | 10 X |
| Content And Storage | Store at 2°C to 8°C. |
| For Use With (Equipment) | 3730xl DNA Analyzer, 3730 DNA Analyzer |
| For Use With (Application) | Standard Sequencing |
| Green Features | Sustainable packaging |
| Product Type | 3730 Running Buffer |
| Quantity | 500 mL |
| Form | Liquid |