Learn More
(S)-(-)-1-Benzyl-3-hydroxypyrrolidine, 99%, ee 99%, Thermo Scientific™
Product Code. 11338856
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11338856
Brand: Thermo Scientific Alfa Aesar L19497.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101385-90-4
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00075485
InChI Key YQMXOIAIYXXXEE-NSHDSACASA-N
Synonym s-1-benzylpyrrolidin-3-ol, s-1-benzyl-3-pyrrolidinol, s-1-benzyl-3-hydroxypyrrolidine, 3s-1-benzylpyrrolidin-3-ol, s-3-hydroxy-1-benzylpyrrolidine, s-1-benzyl-pyrrolidin-3-ol, s---1-benzyl-3-pyrrolidinol, s-1-n-benzyl-3-hydroxypyrrolidine, s-bhp, s-n-benzyl-3-hydroxypyrrolidine
PubChem CID 2733875
IUPAC Name (3S)-1-benzylpyrrolidin-3-ol
SMILES C1CN(CC1O)CC2=CC=CC=C2
Specifications
Density 1.07
Boiling Point 116°C (0.9 mmHg)
Flash Point 148°C (298°F)
Refractive Index 1.548
Quantity 5 g
Beilstein 4982831
Optical Rotation −3.7° (c=5 in Methanol)
Formula Weight 177.25
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (S)-(-)-1-Benzyl-3-hydroxypyrrolidine