missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S)-2-(Boc-amino)butyric acid, 95%

Brand:  Alfa Aesar™ H63686.06

Additional Details : CAS : 34306-42-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410509

  • 138.00€ / 5g


In stock, ready to ship: 1
Add to basket

Description

Description

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

34306-42-8
203.238
PNFVIPIQXAIUAY-LURJTMIESA-N
2755934
CCC(C(=O)O)NC(=O)OC(C)(C)C
C9H17NO4
MFCD00037267
boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

(S)-2-(Boc-amino)butyric acid
34306-42-8
MFCD00037267
boc-abu-oh, boc-l-2-aminobutyric acid, boc-2-abu-oh, s-2-tert-butoxycarbonyl amino butanoic acid, boc-l-2-aminobutanoic acid, boc-l-abu-oh, s-2-tert-butoxycarbonylamino butyric acid, s-2-boc-amino butyric acid, 2s-2-tert-butoxycarbonyl amino butanoic acid, l-2-boc-amino butyric acid
PNFVIPIQXAIUAY-LURJTMIESA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
2755934
Solid
5g
C9H17NO4
6801706
Slightly soluble in water.
CCC(C(=O)O)NC(=O)OC(C)(C)C
203.238
203.24
95%
Videos
SDS Documents