missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Heptanol, 98%, Thermo Scientific Chemicals

Catalog Number p-7050054
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
6033-23-4
C7H16O
116.204
MFCD00065003
CETWDUZRCINIHU-ZETCQYMHSA-N
s-+-2-heptanol, 2s-heptan-2-ol, 2-heptanol, s, unii-7xq9f7kypw, heptan-2s-ol, s-2-heptanol, s-heptan-2-ol, 7xq9f7kypw, d-2-heptanol
2724897
CHEBI:87378
(2S)-heptan-2-ol
CCCCCC(C)O
This item is not returnable. View return policy
6033-23-4
C7H16O
116.204
MFCD00065003
CETWDUZRCINIHU-ZETCQYMHSA-N
s-+-2-heptanol, 2s-heptan-2-ol, 2-heptanol, s, unii-7xq9f7kypw, heptan-2s-ol, s-2-heptanol, s-heptan-2-ol, 7xq9f7kypw, d-2-heptanol
2724897
CHEBI:87378
(2S)-heptan-2-ol
CCCCCC(C)O

(S)-(+)-2-Heptanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.815 g/mL
Boiling Point 160°C to 162°C
Flash Point 59°C (138°F)
Refractive Index 1.421
Quantity 250 mg
UN Number UN1987
Merck Index 14,4662
Solubility Information Soluble in water(3569mg/L at 25°C).
Formula Weight 116.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Heptanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'