Learn More
Branson Ultrasonics™ S-250A Model Sonifier™ Analog Cell Disrupter
Product Code. 10588013
Click to view available options
Wattage:
200 W
Unit Size:
Each
Product Code. 10588013
Brand: Branson Ultrasonics™ 159063674
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ideal device for cell lysis, tissue disruption, and homogenization. Branson Ultrasonics™ S-250A Model Sonifier™ Analog Cell Disrupter has a maximum volume of 500mL.
  • Maximum volume of 500mL
  • Minimum volume of 1mL
  • Analog control
  • 13mm disruptor horn
  • Sound box not included and is a recommended accessory.
Specifications
Wattage 200 W
Capacity (Metric) 500 mL
Description Analog control sonicator/cell disruptor
Includes 13 mm disruptor horn
Weight (Metric) 8.5 kg