missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl-N-[3-(trifluoromethyl)phenylsulfonyl]-DL-homocysteine, 96%, Thermo Scientific™

Catalog Number 15498505
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1009729-55-8
C12H14F3NO4S2
357.362
MFCD06409371
BGORJJLXLJKFMJ-UHFFFAOYSA-N
3-trifluoromethyl phenyl sulfonyl methionine, 4-methylsulfanyl-2-3-trifluoromethyl benzenesulfonamido butanoic acid, 4-methylsulfanyl-2-3-trifluoromethyl-benzenesulfonylamino-butyric acid, 4-methylsulfanyl-2-3-trifluoromethyl phenyl sulfonylamino butanoic acid, n-3-trifluoromethyl phenyl sulfonyl methionine, 4-methylthio-2-3-trifluoromethyl phenyl sulfonyl amino butanoic acid
3485177
4-methylsulfanyl-2-[[3-(trifluoromethyl)phenyl]sulfonylamino]butanoic acid
CSCCC(C(=O)O)NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F
This item is not returnable. View return policy
1009729-55-8
C12H14F3NO4S2
357.362
MFCD06409371
BGORJJLXLJKFMJ-UHFFFAOYSA-N
3-trifluoromethyl phenyl sulfonyl methionine, 4-methylsulfanyl-2-3-trifluoromethyl benzenesulfonamido butanoic acid, 4-methylsulfanyl-2-3-trifluoromethyl-benzenesulfonylamino-butyric acid, 4-methylsulfanyl-2-3-trifluoromethyl phenyl sulfonylamino butanoic acid, n-3-trifluoromethyl phenyl sulfonyl methionine, 4-methylthio-2-3-trifluoromethyl phenyl sulfonyl amino butanoic acid
3485177
4-methylsulfanyl-2-[[3-(trifluoromethyl)phenyl]sulfonylamino]butanoic acid
CSCCC(C(=O)O)NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 107°C to 108°C
Formula Weight 357.37
Percent Purity 96%
Quantity 250 mg
Chemical Name or Material S-Methyl-N-[3-(trifluoromethyl)phenylsulfonyl]-DL-homocysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'