missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Prolinol, 98%, Thermo Scientific Chemicals

Catalog Number p-7045199
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
23356-96-9
C5H11NO
101.149
MFCD00005255
HVVNJUAVDAZWCB-YFKPBYRVSA-N
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
640091
[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)CO
This item is not returnable. View return policy
23356-96-9
C5H11NO
101.149
MFCD00005255
HVVNJUAVDAZWCB-YFKPBYRVSA-N
l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
640091
[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)CO

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water. Soluble in chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Refractive Index 1.4853
Density 1.025
Boiling Point 74°C to 76°C (2mmHg)
Melting Point 42°C to 44°C
Quantity 1 g
Flash Point 86°C (186°F)
Optical Rotation +31° (c=1 in Toluene)
Sensitivity Air sensitive; Hygroscopic
Beilstein 79843
Solubility Information Fully miscible in water. Soluble in chloroform.
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material (S)-(+)-Prolinol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'