missing translation for 'onlineSavingsMsg'
Learn More

(+)-S-Trityl-L-cysteine, 97%, Thermo Scientific Chemicals

Product Code. 11393506
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11393506

Brand: Thermo Scientific Alfa Aesar L14384.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2799-07-7
Molecular Formula C22H21NO2S
Molecular Weight (g/mol) 363.48
MDL Number MFCD00002611
InChI Key DLMYFMLKORXJPO-UHFFFAOYNA-N
Synonym s-trityl-l-cysteine, +-s-trityl-l-cysteine, h-cys trt-oh, s-tritylcysteine, tritylcysteine, 3-tritylthio-l-alanine, 2r-2-amino-3-triphenylmethyl sulfanyl propanoic acid, l-alanine, 3-tritylthio, stlc, s-triphenylmethyl-l-cysteine
PubChem CID 76044
IUPAC Name (2R)-2-amino-3-tritylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)N

Specifications

Melting Point ∽185°C (decomposition)
Beilstein 2339626
Quantity 1 g
Formula Weight 363.48
Optical Rotation +84° (c=1 in Acetic Acid)
Percent Purity 97%
Chemical Name or Material (+)-S-Trityl-L-cysteine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.