missing translation for 'onlineSavingsMsg'
Learn More

(S)-WAY 100135 dihydrochloride, Tocris Bioscience™

Catalog Number 18005832
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
149007-54-5
C24H35Cl2N3O2
468.463
VJGZNBYDSDEOED-FAVHNTAZSA-N
s-n-tert-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-way 100135 dihydrochloride, s-n-t-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, 2s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride
14801907
(2S)-N-tert-butyl-3-[4-(2-methoxyphenyl)piperazin-1-yl]-2-phenylpropanamide;dihydrochloride
CC(C)(C)NC(=O)C(CN1CCN(CC1)C2=CC=CC=C2OC)C3=CC=CC=C3.Cl.Cl
This item is not returnable. View return policy
149007-54-5
C24H35Cl2N3O2
468.463
VJGZNBYDSDEOED-FAVHNTAZSA-N
s-n-tert-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-way 100135 dihydrochloride, s-n-t-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, 2s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride
14801907
(2S)-N-tert-butyl-3-[4-(2-methoxyphenyl)piperazin-1-yl]-2-phenylpropanamide;dihydrochloride
CC(C)(C)NC(=O)C(CN1CCN(CC1)C2=CC=CC=C2OC)C3=CC=CC=C3.Cl.Cl

Potent and selective 5-HT1A antagonist

Potent, selective 5-HT1A receptor antagonist (IC50 = 15 nM). Selective over 5-HT1B, 1C, 2,α1, α2 and D2 receptors (IC50 > 1000 nM). Centrally active on systemic administration.
Quantity 50 mg
Formula Weight 468.47
Percent Purity >99%
Chemical Name or Material (S)-WAY 100135 dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'