Learn More
S.C.A.T.™ S 60/61 Safety Waste Caps
Chemically resistant safety waste cap with accessory connector options including vapor filter capacity. S.C.A.T.™ S 60/61 Safety Waste Caps are constructed of pure PTFE and PE-HD.
Marque:  S.C.A.T.™ 107916
Informations supplémentaires : Poids : 0.01000kg
Code produit. 10100050
  • 149.66€ / 1 pièce
nous consulter 056 260 260
Ajouter au panier
Description
Description
  • Variety of accessory connectors available
  • Different thread sizes available
  • Accessories fit onto your available containers, or onto orderable containers
  • Resistant against aggresive media
  • Resistant against organic solvents, acids and lyes
Spécification
Spécification
Includes 3 capillary connectors (2.3/3.2 mmO.D.), connector for exhaust filter, ground connection, S 60/61 thread
Waste container
PTFE, HDPE
Safety Waste Cap
Vidéos