missing translation for 'onlineSavingsMsg'
Learn More

Melford Sabouraud Dextrose Agar

Product Code. 30018768
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018768

Brand: Melford S12500500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sabouraud Dextrose Agar is an agar medium recommended for the isolation, cultivation and identification of non-pathogenic and pathogenic fungi.

Sabouraud Dextrose Agar is an agar medium recommended for the isolation, cultivation and identification of non-pathogenic and pathogenic fungi. This medium does not contain antibiotics or inhibitors and can be used for the cultivation of those fungi which are susceptible to antibiotics.

Dissolve 65.0 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Casein Digest Peptone: 10.0 g/L

Dextrose: 40.0 g/L

Agar: 15.0 g/L

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.