missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Sabouraud Dextrose Agar, EP/USP

Sabouraud Dextrose Agar (SDA) is designed for determination of yeasts and molds and the examination of non-sterile pharmaceutical products. Prepared according to European, Japanese and United States Pharmacopoeia recommendations; packaged in 200mL screw-top bottles or 18mL capped tubes.

Brand:  Merck Millipore 1463930006

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12738818

  • 46.21€ / 200mL


Estimated Shipment: 07-07-2021
Add to basket

Description

Description

Sabouraud Dextrose (4%) Agar (SDA) is a complex medium for cultivation and isolation of yeasts and molds. The media is also used for microbiological examination of non-sterile products. The high concentration of Dextrose and low pH promote yeast/mold growth while inhibiting bacterial growth. The media also promotes the formation of spores (conidia and sporangia) and pigments.

  • Composition (per L): Casein Peptone 5 g; Meat Peptone 5 g; Dextrose 40 g; Agar 15 g
  • Prepared according to the recommendations of the current European, Japanese and United States Pharmacopoeia
  • Used in microbial examination of non-sterile pharmaceutical products
  • Designed for the determination of the total count of yeasts and molds
  • Available in different filling volumes:
    • 25mL tubes with yellow caps, Fill volume: 18mL
    • 250mL bottle with screw caps, Fill volume: 200mL
Specifications

Specifications

Screw cap
Yellow
20 - 25 °C
Sabouraud Dextrose Agar (SDA)
50 - 200%
200 mL
6 Bottles
European, Japanese and United States Pharmacopoeia (EP, 2.6.13.; JP, 4.05 and USP, 62)
Blue
Per 1L: Casein Peptone 5 g; Meat Peptone 5 g; Dextrose 40 g; Agar 15 g
Ready for use
Store upright, protected from light and properly sealed at +2 °C to +25 °C
Determination of the total count of yeasts and molds
Yeast
Bottle
250 mL Bottle
Videos
SDS
Documents
Certificates