Learn More
Sartorius Stedim Biotech™ Sabouraud NKS NPS and Gridded e.motion MF
Product Code. p-7084296 Shop All Sartorius Products
Click to view available options
Format:
47 mm Disc
50 mm Disc
Unit Size:
Each
Pack of 100
Product Code. 15972490
Brand: Sartorius Stedim Biotech™ 1406947RDN
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes dehydrated media pads in petri dishes , pre-sterilized, along with suitable membrane filters for economical, time-saving microbiological quality control.
  • Nutrient pads with gridded membrane filters
  • For determination of yeasts and molds
  • Trouble-free storage up to 24 months
  • Compliance with international standards and references
  • Validation guide available
  • Highly versatile - multiple applications
  • Economical by time- and cost-savings
  • Easy handling
  • Dispenser ready membrane filters designed for use with Microsart&trade: e.motion dispenser
TRUSTED_SUSTAINABILITY
Specifications
Format 47 mm Disc
Media Type Sabouraud NKS
Sterility Sterile
Target Organism Class Yeast, Molds
Quantity 100/Pk.
Diameter (Metric) 47 mm