Learn More
WLD Tec™ Safety Adapter 360
Description
- For Flame 100.
- With integrated pressure regulator
- For cartridges CV 360, 52 g from Camping Gaz.
- Continuous cartridge operation 65 minutes.
Specifications
Specifications
| For Use With (Equipment) | Flame100 |