Learn More
Justrite™ Safety Cabinet Piggyback
Brand:  Justrite™ 8917221
Additional Details : Weight : 68.49244kg
Product Code. 11935426
  • 2070.00€ / Each
Estimated Shipment: 02-10-2024
to see stock.
Description
Description
2 door, self close, not recommended for phenol, nitric/sulphuric acid steel, designed to mount on top of standard 114/170L cabinet blue 610mm x 1,092mm x 457mm (w x h x d) 64L