missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT™ Safety Canisters, HDPE-EC
Click to view available options
Closure Size:
S50
S60/61
S70/71
S90
Includes:
Canister
Canister with Blue Stripes on Grip, without UN-Y Approval
Canister with Floater
Canister with Green Stripes on Grip, without UN-Y Approval
Canister with Red Stripes on Grip, without UN-Y Approval
Canister with UN-Y Approval
Canister with Viewing Strip, UN-Y Approval
Canister with Yellow Stripes on Grip, without UN-Y Approval
Floater
UN-Y approval
Capacity (Metric):
5 L
10 L
20 L
30 L
60 L
Description
- Made of very robust and electrically conductive HDPE, which is very resistant to solvents commonly used in laboratories
- Filling quantities from 10 L for use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g. professional SymLine™ disposal systems)
- Thread types S50, S60/61, S70/71 and S90 are commonly used in the laboratory and are all compatible
- Safety Waste Caps range for the safe disposal of volatile, hazardous substances
- Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
- Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets
Specifications
Product Type | Canister |
Closure Size | S60/61 |
Capacity (Metric) | 30 L |
Dimensions (L x W x H) | 240 x 364 x 455 mm |
Material | HDPE-EC |
For Use With (Application) | Collection of Flammable Liquid Waste |
Includes | Canister with Floater |
Autoclavable | Not Autoclavable |
Color | Black |
For Use With (Equipment) | SCAT™ Safety Funnels |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'