Learn More
Lenz Laborglasintrumente™ “Safety Clip” Conical Joint Clips
POM safety clip for conical ground joints
35.72€ - 46.16€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11933361
|
Lenz Laborglasintrumente™
01103824 |
For ground joint NS 24/29 |
35.72€
Pack of 10 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11963361
|
Lenz Laborglasintrumente™
01103845 |
For ground joint NS 45/40 |
46.16€
Pack of 10 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- Height adjustable by variable gear rods, also for use with sleeves with grip collar
- Outstanding chemical resistance
- Operating temperature up to approximately 150 °C
Specifications
10 | |
POM | |
Blue |