Learn More
RETSCH Safety Closure Devices
1015.00€ - 1580.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15137696
|
RETSCH
22.867.0007 |
For 80 mL Agate Comfort Grinding Jars |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
10680822
|
RETSCH
22.867.0003 |
For 80 mL, 125 mL Stainless Steel or Tungsten Carbite Comfort Grinding Jars |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
10651392
|
RETSCH
22.867.0004 |
For Comfort 250 mL Grinding Jars |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
10074831
|
RETSCH
22.867.0005 |
For Comfort 500 mL Grinding Jars |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
10564102
|
RETSCH
22.867.0002 |
For Comfort 50 mL Grinding Jars |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
Safety Closure Device |