missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Safety Equipment Zero Waste Box™
Recycle Safety Equipment using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
136.09€ - 316.79€
Specifications
Color | Beige, Green, Red, White |
---|---|
Disposable | Single-use |
Latex Free | Yes |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17855705
|
Terracycle™
CS58-S-BE |
Small |
136.09€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17865965
|
Terracycle™
CS58-M-BE |
Medium |
209.15€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17805895
|
Terracycle™
CS58-L-BE |
Large |
316.79€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
Use this Zero Waste Box to recycle:
- Earplugs
- Beard nets
- Hairnets
- Disposable gloves
- Safety glasses
- Disposable garments
- Disposable shoe covers
- Disposable masks*
*We accept all types of disposable face masks made from plastic, such as dust masks, FFP2 (or so-called ''N95''), FFP3 and surgical masks (as long as they don't come from a medical environment).
Do not send in:
- Medical waste
- This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Autoclaved material
- Fabric, homemade or paper face masks
- Wax earplugs
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which present a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass
Specifications
Beige, Green, Red, White | |
Yes | |
Not Autoclavable | |
Recycling Box |
Single-use | |
Non-sterile | |
Cardboard, Plastic |