Learn More
Uvex Safety Gloves Phynomic 60068
Product Code. p-200105925 Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
Product Code. 30245729
Brand: Uvex 6006806
Only null left
Add to basket
Add to basket
Phynomic assembly and cut protection gloves are especially ergonomically shaped, sit like a second skin and are very light, flexible and breathable.
  • Free of accelerators and dermatologically tested for skin compatibility (proDERM Institute)
  • Ideal for allergy sufferers
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Aqua-Polymer
Color Black
Cut Resistance Rating Level F
Hand Ambidextrous
Latex Free Yes
Material Elastane, HPPE, Polyamide, Steel Fiber
Product Type Mechanical/Cut Protection Gloves (Heavy-duty)
Size 6