missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ KleenGuard™ Jackson Safety™ V30 Nemesis™ Eyewear

Catalog Number 12661406
Click to view available options
Lens Tint: IR/UV 5.0
This item is not returnable. View return policy
This item is not returnable. View return policy

Kimberly-Clark™ JACKSON SAFETY™ V30 Nemesis™ Eyewear offers protection from sparks and small debris while working in welding or related areas.

  • Complies with: EN166:2001 1F for safety
  • Complies with: EN166:2001 level F for impact resistance
  • Complies with: EN166:2001 class 1 for optical clarity
  • 99% UVA/UVB protection
  • Comfortable to wear for long periods
  • Economical eye protection for workers and visitors
TRUSTED_SUSTAINABILITY
Certifications/Compliance EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity.
Lens Tint IR/UV 5.0
Product Type Safety Eyewear
For Use With (Application) Protecting eyes from sparks, spatter and small debris while working in welding or related areas, driving compliance through appealing styling
No. per Pack 12/Pk.
Temple Style Soft touch
Weight (Metric) 0.324 kg
Frame Type Wrap-around, IRUV 5.0 LENS
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'