Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
85.75€ - 710.00€
Specifications
Type | Safety Waste Cap |
---|---|
For Use With (Application) | Waste Disposal |
For Use With (Equipment) | SymLine System |
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | ||||
17145501
|
SymLine™
306497 |
GL45 | 2 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Tube Connector for (ID) 5 to 11.5 mm, Collector for HPLC Waste |
95.30€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
17135501
|
SymLine™
306491 |
GL45 | 3 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Collector for HPLC Waste |
85.75€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
16876011
|
SymLine™
306494 |
GL45 | 5 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Collector for HPLC Waste |
112.00€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
15960386
|
SymLine™
106478 |
S55 | Electronic Level Control, GL25 (m), Immersed Filling Lance, Connector for Exhaust Filter |
388.00€
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
15724922
|
SymLine™
106522 |
S55 | 1 x GL25 (m), Immersed Filling Lance, Connector for Exhaust Filter |
178.00€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17808743
|
SymLine™
106719 |
S55 | Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter |
560.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17079693
|
SymLine™
306581 |
S60/61 | Adjustable PTFE Level Control, Cable Connection (2 m), Connector for Exhaust Filter |
585.00€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
16866011
|
SymLine™
306482 |
S60/61 | 2 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Tube Connector for (ID) 5 to 11.5 mm |
252.00€
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
17813352
|
SymLine™
106718 |
S60/61 | Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter |
560.00€
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
17377409
|
SymLine™
106484 |
S90 | Electronic Level Control, GL25 (m), Connector for Exhaust Filter |
510.00€
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
17838743
|
SymLine™
106720 |
S90 | Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter |
710.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Safety Waste Cap | |
Waste Disposal | |
SymLine System |