missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SAG

Product Code. p-7217761
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17110222

Brand: STEMCELL Technologies 73412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hedgehog pathway activator; Activates Smoothened (SMO)

  • SAG (Smoothened Agonist) is a chlorobenzothiophene-containing compound which acts as an activator of the G protein-coupled receptor Smoothened (SMO, EC50 = 3 nM; Chen et al.) SMO is a component of the Hedgehog signaling pathway, which is translocated to the primary cilium after stimulation of the Patched receptor by Hedgehog family ligands, leading to pathway activation
  • SAG activates SMO via direct binding to the heptahelical bundle (Kd= 59 nM), stabilizing a specific conformation of SMO in cilia and leading to increased downstream gene expression (Rohatgi et al.) SAG abrogates cyclopamine inhibition of SMO, indicating that it acts downstream of cyclopamine (Frank-Kamenetsky et al.; Chen et al.; Lewis & Krieg)

Specifications

CAS 912545-86-9
Synonym Smoothened Agonist
Quantity 1 mg
Molecular Formula C28H28ClN3OS
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.