missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SAHA

Product Code. 17110292
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17110292 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17110292 Supplier STEMCELL Technologies Supplier No. 73902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epigenetic modifier; Inhibits HDAC1 and HDAC3

  • SAHA (suberoylanilide hydroxamic acid) is a potent inhibitor of class I and class II histone deacetylases (HDACs)
  • This compound inhibits the activities of HDAC1 and HDAC3 (IC50 = 10 nM and 20 nM, respectively; Richon et al.) SAHA exhibits antiproliferative properties and can cause cell cycle arrest at G1 (Marks & Breslow)
  • Improves reprogramming efficiency from human and mouse somatic to induced pluripotent stem cells (Huangfu et al.)
  • Downregulates homologous recombination and leads to apoptosis in ovarian cancer cells when used in combination with olaparib (Konstantinopoulos et al.)
  • SAHA downregulates Nanog expression, which leads to proliferation inhibition in head and neck cancer stem cells (Kumar et al.)

Specifications

CAS 149647-78-9
Synonym Suberoylanilide hydroxamic acid; Vorinostat
Quantity 100 mg
Molecular Formula C14H20N
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.