Learn More
Saint Gobain Life Sciences Chemware™ PTFE PFA Griffin Beakers
Click to view available options
Capacity (Metric):
50 mL
125 mL
Description
- Designed for rapid heat transfer
- Can be heated in a microwave or directly on a hot plate up to 500°F (260°C)
- Transparent
- Graduated for easy measuring
- Autoclavable
Specifications
Capacity (English) | 1.7 oz. |
Color | Transparent |
Capacity (Metric) | 50 mL |
Material | PTFE |
Graduation Range | 10 to 50 mL |
Type | Beaker |
Autoclavable | Autoclavable |
Disposable | Reusable |
Graduated | Yes |
Style | Griffin |