Learn More
Saline-Sodium Phosphate-EDTA (SSPE), 20X Solution, Fisher BioReagents
Product Code. 10499113
Click to view available options
Quantity:
1L
Unit Size:
1 L
Product Code. 10499113
Brand: Fisher BioReagents BP13281
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Saline–Sodium Phosphate–EDTA (SSPE) is used in bacterial screening and hybridization procedures.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 2.9 to 3.1M
Form Liquid
Quantity 1L
Storage Requirements RT