Learn More
Masterflex™ Anti-Siphon Valves
Product Code. 13513010
Click to view available options
Material:
SAN
Unit Size:
Pack of 10
Product Code. 13513010
Brand: Masterflex™ 3050591
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Connect these valves to bio-bags or sampling tubes. Cole-Parmer™ Anti-Siphon Valves are molded onto the tubing for a seamless transition.
Valves feature low priming volume, ease of removing air bubbles during priming and operate in any spatial orientation. Manufactured DEHP-and latex-free silicone diaphragms. Materials meet USP CLass VI and ISO 10993 specifications. Each assembly comes packaged individually and can be sterilized..
Specifications
Material SAN
Packaging Quantity 10 Pack
Product Type Luer Fitting