Learn More
Skytec™ Sapphire Aero Gloves
Product Code. 30274579
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274579
Brand: Skytec™ SKG00011FD
Only null left
Add to basket
Add to basket
A second skin cut resistant glove, thin, soft and comfortable allowing high dexterity tasks.
  • Ultra-Lightweight
  • Designed for easy movement and continuous wear
Specifications
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A3, EN 388:2016 + A1:2018 4X42C, EN 21420:2020
Construction Seamless Knit
Coating Material Foam Nitrile
Color Gray, Black
Cut Resistance Rating Level C
Hand Ambidextrous
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size Small
EN388 Standard 4X42C
Length (Metric) 220 mm