Learn More
Skytec™ Sapphire Total Gloves
Product Code. 30274554
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. Size unitSize
30274554 Large 10 pairs
30274529 Small 10 pairs
30274552 Medium 10 pairs
30274512 X-Large 10 pairs
30274616 2X-Large 10 pairs
5 options
Product Code. 30274554
Brand: Skytec™ SKY00123
Only null left
Add to basket
Add to basket
A great fitting, fully coated glove combining high cut protection and excellent grip in oily and wet conditions. Ideal for oily tasks with sharp parts that require great grip.
  • Oil Resistant Water Repellent Cut Resistant Grip Glove
  • Cut resistant: Level E/ ANSI A5
  • A flexible
  • No irritation by seams in contact with the skin
  • Elasticated wrist provides a secure fit
Specifications
Certifications/Compliance (Europe) CE II, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 + A1:2018 4X43E, EN 407:2020 X1XXXX, EN 21420:2020
Construction Seamless Knit
Coating Material Foam Nitrile
Color Black
Cut Resistance Rating Level E
Hand Ambidextrous
Latex Free Yes
Material Steel Fiber
Product Type Cut-resistant Gloves
Size Large
EN388 Standard 4X43E
Liner Material HPPE, Nylon, Spandex
Length (Metric) 250 mm