Learn More
Skytec™ Sapphire Zinc Gloves
Product Code. 30274612
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274612
Brand: Skytec™ SKG00086FJ
Only null left
Add to basket
Add to basket
Glass and steel fiber free uncoated gloves offering high cut protection. Can be used as an under glove or worn by itself.
  • High Cut Resistant Uncoated Glove Made with Engineered HPPE
  • Cut resistant: Level E / A9
  • Engineered HPPE offers outstanding cut protection
  • Glass and steel fiber free reduce risks of irritation
  • Spandex in the liner allows the glove to fit the hand perfectly
  • Designed for easy movement and continuous wear
  • Breathable to reduce perspiration
  • Seamless knit prevents irritation
  • Elasticated wrist provides a secure fit
  • Ambidextrous for easy and quick donning
Specifications
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 + A1:2018 2X43E, EN 21420:2020
Coating Material Uncoated
Color Gray
Cut Resistance Rating Level E
Hand Ambidextrous
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard 2X43E
Length (Metric) 260 mm