missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SARS-CoV-2 NSP6 (YP_009725302.1) VersaClone cDNA

Product Code. 18704197
Change view
Click to view available options
:
10 μg
Product Code.
18704197 10 μg
1 options
This item is not returnable. View return policy

Product Code. 18704197

Brand: R&D Systems™ RDC3154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene NSP6
Accession Number YP_009725302.1
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 889 bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species SARS-CoV-2
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.