Learn More
IBA Lifesciences SARS-CoV-2 Peptide PYRVVVLSF (HLA-A*2402)
GREENER_CHOICE
Product Code. 16634754 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 16634754
Brand: IBA Lifesciences 67122901
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Single peptide synthesized as presented by the MHC class I allele
  • SARS-CoV-2 peptide PYRVVVLSF (HLA-A*2402) is a single peptide for stimulation of T cells.
  • The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*2402 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Purity >85%
Endotoxin Level Free
Format Lyophilized Powder
Quantity 1 mg
Research Category T cell stimulation