missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Sartopore™ 2 SartoScale Polyethersulfone Membrane Flat Disc Capsules
Brand: Sartorius™ 5445307GS--FF--M
Additional Details : Weight : 0.50000kg
Specifications
Autoclavable | |
PES | |
Sterile | |
0.25 in. Multiple Stepped Hose Barb | |
0.25 in. Multiple Stepped Hose Barb | |
Flat Disc Capsule |
ISO 9001, USP Class VI, cGMP Standard | |
0.2 μm | |
17.3 m2 | |
1Each | |
1 Each |