missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Sartopure™ Filter, Glass Fiber

Optimized for clarification and retention via multiple layers of progressively finer adsorptive glass fleece material.

Brand:  Sartorius Stedim Biotech™ 5555305P0--SS--V

Additional Details : Weight : 0.10000kg

Product Code. 15963630

  • 556.50€ / Pack of 2
Estimated Shipment: 12-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Turbidity reduction results in optimal protection of all downstream material like chromatographic columns or membrane filters.
  • Turbidity reduction in cell harvest, mAB, vaccine, lipid- and colloid-containing fluids.
  • Highly adsorptive.
Specifications

Specifications

Autoclavable
0.25 m2
50 mm
2 bar
330 mm
Silicone O-ring/Seal, Glass Fiber Prefilter
Polypropylene
Tri-Clamp
Tri-Clamp
108.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions