Learn More
Sartorius Qualitative Grade 1291 Filter Papers
Product Code. 10012411 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
55 mm
70 mm
90 mm
110 mm
125 mm
150 mm
185 mm
240 mm
Format:
Disc
Unit Size:
Pack of 100
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Format Unit Size
10012411 55 mm Disc Pack of 100
10052431 125 mm Disc Pack of 100
10585891 90 mm Disc Pack of 100
10767432 240 mm Disc Pack of 100
10674522 185 mm Disc Pack of 100
10682582 70 mm Disc Pack of 100
10103901 110 mm Disc Pack of 100
10377401 150 mm Disc Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10012411 Supplier Sartorius Supplier No. FT3209055
 Restricted Item
In Stock
Add to basket
Add to basket
These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration.
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content < 0,1% according to DIN 54370. Wet-strengthened.
TRUSTED_SUSTAINABILITY
Specifications
Format Disc
Material Cotton Linters, Refined Pulp
Particle Retention 2 to 3 μm
Pore Size 2 to 3 μm
Filtration Speed 180 sec.
Grade 1291
Quantity 100/Pk.
Thickness (Metric) 0.16 mm
Type Qualitative Filter Paper
Diameter (Metric) 55 mm
For Use With (Equipment) Conical Funnel