Learn More
Sartorius Qualitative Grade 292 a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand: Sartorius FT-3-215-320
8.86 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.81000kg
Description
- Pure cotton linters or cotton linters with refined pulp
- No additives, such as wet-strengthening agents
Specifications
Disc | |
97 sec. | |
100/Pk. | |
320 mm |
4 to 7 μm | |
292 a | |
0.18 mm |